Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

719773

Sigma-Aldrich

2,2,4-Trimethyl-1,3-pentanediol-1,3-13C2 monoisobutyrate-1-13C

mixture of isomers, 99 atom % 13C, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CH13CH[O(R)]C(CH3)213CH2O(R), R=-13COCH(CH3)2 or H
Molecular Weight:
219.30
UNSPSC Code:
12352001
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C

assay

97% (CP)

bp

255 °C (lit.)

mp

-50 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

mass shift

M+3

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service