Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

718688

Sigma-Aldrich

Bis(tert-butylimino)bis(tert-butylamino)tungsten

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H9NH)2W(C4H9N)2
CAS Number:
Molecular Weight:
470.34
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

99.9% trace metals basis

form

powder or crystals
solid

reaction suitability

core: tungsten

mp

78-91 °C

SMILES string

CC(C)(C)N[W](NC(C)(C)C)(=NC(C)(C)C)=NC(C)(C)C

InChI

1S/2C4H10N.2C4H9N.W/c4*1-4(2,3)5;/h2*5H,1-3H3;2*1-3H3;/q2*-1;;;+2

InChI key

SJPFZRACRCONRE-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Flam. Sol. 2 - Skin Corr. 1B - Water-react 2

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service