Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

718262

Sigma-Aldrich

Benzene-D6

99.6 atom % D

Synonym(s):

Hexadeuterobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15
Beilstein/REAXYS Number:
1905426
EC Number:
MDL number:
UNSPSC Code:
12142201

isotopic purity

99.6 atom % D

assay

≥99.00%

form

liquid

expl. lim.

0.34-6.3 % (lit.)

impurities

≤0.0100% water
water

refractive index

n20/D 1.497 (lit.)

bp

79.1 °C/760 mmHg (lit.)

mp

6.8 °C (lit.)

density

0.950 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

UHOVQNZJYSORNB-MZWXYZOWSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

target_organs

Blood

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

12.2 °F - closed cup

flash_point_c

-11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyong Lu et al.
Molecules (Basel, Switzerland), 19(9), 14292-14303 (2014-09-13)
Gated molecular encapsulations, with baskets of type 1, are postulated to occur by the mechanism in which solvent molecule penetrates the inner space of 1, through one of its apertures, while the residing guest simultaneously departs the cavity. In the
Zachariah M Norman et al.
ACS nano, 8(7), 7513-7521 (2014-06-25)
We report the evolution of electrical transport and grain size during the sintering of thin films spin-cast from soluble phosphine and amine-bound, chloride-terminated cadmium selenide nanocrystals. Sintering of the nanocrystals occurs in three distinct stages as the annealing temperature is
Alejandro G Lichtscheidl et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16156-16163 (2015-08-22)
The synthesis and full characterization, including Nuclear Magnetic Resonance (NMR) data ((1)H, (13)C{(1)H} and (119)Sn{(1)H}), for a series of Me3SnX (X = O-2,6-(t)Bu2C6H3 (), (Me3Sn)N(2,6-(i)Pr2C6H3) (), NH-2,4,6-(t)Bu3C6H2 (), N(SiMe3)2 (), NEt2, C5Me5 (), Cl, Br, I, and SnMe3) compounds in
Ursula J Williams et al.
Dalton transactions (Cambridge, England : 2003), 43(43), 16197-16206 (2014-08-26)
The trivalent compound K[Ce[N(SiHMe2)2]4] was synthesized and oxidized, providing a convenient route to the reported cerium(IV) compound Ce[N(SiHMe2)2]4. Protonolysis reactions of Ce[N(SiHMe2)2]4 with tert-butanol, substituted benzyl alcohols, and 2,6-diphenylphenol yielded the neutral tetravalent compounds Ce(O(t)Bu)4(py)2, Ce2(OCH2C6R5)8(thf)2 (R = Me, F)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service