Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

717622

Sigma-Aldrich

2-Formyl-4,5-methylenedioxyphenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7BO5
CAS Number:
Molecular Weight:
193.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

form

powder

functional group

aldehyde

SMILES string

OB(O)c1cc2OCOc2cc1C=O

InChI

1S/C8H7BO5/c10-3-5-1-7-8(14-4-13-7)2-6(5)9(11)12/h1-3,11-12H,4H2

InChI key

AKEPUIJBVDGLMX-UHFFFAOYSA-N

Application

Reactant for:
  • Enantioselective preparation of indenamines by cationic palladium complex-catalyzed tandem annulation with alkynes
  • Stereoselective preparation of indenol derivatives via cationic palladium complex-catalyzed diastereoselective tandem annulation
  • Synthesis of oxazoline-substituted potassium organotrifluoroborates
  • Preparation of aristolactam analogs as an antitumor agent
  • Suzuki-Miyaura coupling reactions
  • Suzuki and Negishi cross-coupling reactions

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service