716529
2-Methylindole-7-carboxaldehyde
97%
Synonym(s):
2-Methyl-1H-indole-7-carbaldehyde, 2-Methyl-1H-indole-7-carboxaldehyde, 2-Methyl-7-formylindole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H9NO
CAS Number:
Molecular Weight:
159.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
form
solid
mp
72-76 °C
SMILES string
Cc1cc2cccc(C=O)c2[nH]1
InChI
1S/C10H9NO/c1-7-5-8-3-2-4-9(6-12)10(8)11-7/h2-6,11H,1H3
InChI key
KMTJSJFOHDJFCF-UHFFFAOYSA-N
Application
Reactant for preparation of:
- Chiral indole-based chromium(III) complexes as enantioselective catalysts for the conversion of propylene oxide and carbon monoxide to enantioenriched β-butyrolactone
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service