Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

716081

Sigma-Aldrich

Methyl-13C phenyl sulfide

99 atom % 13C

Synonym(s):

Methyl-13C phenyl thioether, Thioanisole-(methyl-13C)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$461.00

$461.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$461.00

About This Item

Linear Formula:
C6H5S13CH3
CAS Number:
Molecular Weight:
125.20
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.12

$461.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

form

liquid

bp

188 °C (lit.)

density

1.065 g/mL at 25 °C

mass shift

M+1

SMILES string

[13CH3]Sc1ccccc1

InChI

1S/C7H8S/c1-8-7-5-3-2-4-6-7/h2-6H,1H3/i1+1

InChI key

HNKJADCVZUBCPG-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

Licensed from Highlands Stable Isotopes Corp.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

163.4 °F - closed cup

flash_point_c

73 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service