Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

716073

Sigma-Aldrich

1,3-Dithiane-2-13C-2,2-d2

98 atom % D, 99 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,980.00

$1,980.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$1,980.00

About This Item

Empirical Formula (Hill Notation):
13C2C2D2H6S2
Molecular Weight:
124.23
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

$1,980.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C
98 atom % D

Quality Level

form

solid

mp

52-54 °C

mass shift

M+3

SMILES string

[2H][13C]1([2H])SCCCS1

InChI

1S/C4H8S2/c1-2-5-4-6-3-1/h1-4H2/i4+1D2

InChI key

WQADWIOXOXRPLN-RCZUCQLYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

Licensed from Highlands Stable Isotopes Corp.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service