Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

715999

Sigma-Aldrich

Diethyl acetamidomalonate-1,2,3-13C3

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONH13CH(13CO2C2H5)2
Molecular Weight:
220.20
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C

assay

98% (CP)

form

solid

mp

95-98 °C (lit.)

mass shift

M+3

SMILES string

CCO[13C](=O)[13CH](NC(C)=O)[13C](=O)OCC

InChI

1S/C9H15NO5/c1-4-14-8(12)7(10-6(3)11)9(13)15-5-2/h7H,4-5H2,1-3H3,(H,10,11)/i7+1,8+1,9+1

InChI key

ISOLMABRZPQKOV-ULEDQSHZSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service