Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

715867

Sigma-Aldrich

Phenyl vinyl-2-13C sulfone

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH=13CH2
CAS Number:
Molecular Weight:
169.21
MDL number:
UNSPSC Code:
12352005

isotopic purity

99 atom % 13C

form

solid

mp

67-69 °C

mass shift

M+1

SMILES string

[13CH2]=CS(=O)(=O)c1ccccc1

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

Licensed from Highlands Stable Isotopes Corp.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service