715743
3′-Bromo-2,2,2-trifluoroacetophenone
97%
Synonym(s):
3-Bromo-α,α,α-trifluoroacetophenone
About This Item
assay
97%
form
liquid
refractive index
n20/D 1.504
density
1.637 g/mL at 25 °C
Storage temp.
2-8°C
SMILES string
FC(F)(F)C(=O)c1cccc(Br)c1
InChI
1S/C8H4BrF3O/c9-6-3-1-2-5(4-6)7(13)8(10,11)12/h1-4H
Inchi Key
JOKNUXPHMQZTQB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Rieke is a registered trademark of Rieke Metals, Inc.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service