Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

715344

Sigma-Aldrich

4-Bromo-1-isobutylpyrazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11BrN2
CAS Number:
Molecular Weight:
203.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

density

1.332 g/mL at 25 °C

functional group

bromo

SMILES string

CC(C)Cn1cc(Br)cn1

InChI

1S/C7H11BrN2/c1-6(2)4-10-5-7(8)3-9-10/h3,5-6H,4H2,1-2H3

InChI key

HDXYNFLYDLDONH-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

219.0 °F - closed cup

flash_point_c

103.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service