Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71477

Sigma-Aldrich

Phosphazene base P2-t-Bu on polystyrene

extent of labeling: ~1.6 mmol/g loading

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

form

beads

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

~1.6 mmol/g loading

matrix

crosslinked with 2% DVB

particle size

100-200 mesh

Pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service