Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

714496

Sigma-Aldrich

4-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3ClF4O2S
CAS Number:
Molecular Weight:
262.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

48-52 °C

SMILES string

Fc1ccc(c(c1)C(F)(F)F)S(Cl)(=O)=O

InChI

1S/C7H3ClF4O2S/c8-15(13,14)6-2-1-4(9)3-5(6)7(10,11)12/h1-3H

Inchi Key

IGMYEVQPXWKFQF-UHFFFAOYSA-N

Pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service