Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

713406

Sigma-Aldrich

1-tert-Butyl-4-piperidone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO
CAS Number:
Molecular Weight:
155.24
Beilstein/REAXYS Number:
471469
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥96.5% (GC)
96.5-103.5% (T)
97%

form

liquid

SMILES string

CC(C)(C)N1CCC(=O)CC1

InChI

1S/C9H17NO/c1-9(2,3)10-6-4-8(11)5-7-10/h4-7H2,1-3H3

InChI key

NKPMJJGLSFUOPK-UHFFFAOYSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service