Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

713368

Sigma-Aldrich

(S)-α-(Phenylmethyl)-1-pyrrolidinethanamine

96%

Synonym(s):

(S)-2-Phenyl-1-pyrrolidin-1-ylmethyl-ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20N2
CAS Number:
Molecular Weight:
204.31
Beilstein/REAXYS Number:
10160096
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥95.5% (GC)
95.5-104.5% (NT)
96%

form

liquid

optical activity

[α]/D +5.0±1.0°, c = 1 in methanol

functional group

amine
phenyl

storage temp.

2-8°C

SMILES string

N[C@H](CN1CCCC1)Cc2ccccc2

InChI

1S/C13H20N2/c14-13(11-15-8-4-5-9-15)10-12-6-2-1-3-7-12/h1-3,6-7,13H,4-5,8-11,14H2/t13-/m0/s1

InChI key

YDLCAQYNXGDYTM-ZDUSSCGKSA-N

Application

(S)-α-(Phenylmethyl)-1-pyrrolidinethanamine can be used as an organocatalyst:
  • In the Michael addition reaction of α-substituted vinyl ketones with malononitriles.[1]
  • In the aldol reactions of α-hydroxyketones with substituted aldehydes.[2]
  • In the asymmetric conjugate addition of vinyl ketones to azoles.[3]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral primary amine catalysed asymmetric conjugate addition of azoles to α-substituted vinyl ketones
Fu N, et al.
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 1(1), 68-72 (2014)
Chiral primary amine catalyzed asymmetric Michael addition of malononitrile to α-substituted vinyl ketone
Fu, Niankai and Zhang, Long and Luo, Sanzhong
Organic Letters, 17(2), 382-385 (2015)
Chiral Primary- Tertiary Diamine Catalysts Derived From Natural Amino Acids for syn-Aldol Reactions of Hydroxy Ketones
Li J, et al.
The Journal of Organic Chemistry, 74(4), 1747-1750 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service