Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

713341

Sigma-Aldrich

4-Nitrophenyl N-benzylcarbamate

≥98.0%

Synonym(s):

Benzylcarbamic acid 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2O4
CAS Number:
Molecular Weight:
272.26
Beilstein/REAXYS Number:
4261736
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (HPLC)
≥98.0%

form

powder

SMILES string

[O-][N+](=O)c1ccc(OC(=O)NCc2ccccc2)cc1

InChI

1S/C14H12N2O4/c17-14(15-10-11-4-2-1-3-5-11)20-13-8-6-12(7-9-13)16(18)19/h1-9H,10H2,(H,15,17)

InChI key

OCAOEODBVXZHNZ-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Radiofluorinated quinoxalinedione derivatives as potential ligands of the AMPA receptor
  • Mono-substituted urea side chains in solid phase peptide synthesis
  • 4-substituted-1,2,4-triazolidine-3,5-diones
  • Aryl carbamates as probes for the inhibition mechanism of cholesterol esterase
Reagent for the conversion of amine into monosubstituted ureas.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service