Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

713120

Sigma-Aldrich

1-Butyl-1-methylpyrrolidinium hexafluorophosphate

≥97.5% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20F6NP
CAS Number:
Molecular Weight:
287.23
Beilstein/REAXYS Number:
11148397
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.5% (T)

form

powder

impurities

≤0.5% water

SMILES string

F[P-](F)(F)(F)(F)F.CCCC[N+]1(C)CCCC1

InChI

1S/C9H20N.F6P/c1-3-4-7-10(2)8-5-6-9-10;1-7(2,3,4,5)6/h3-9H2,1-2H3;/q+1;-1

InChI key

HCGXEKKFZYYPFF-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service