Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

713082

Sigma-Aldrich

1-Butyl-1-methylpiperidinium tetrafluoroborate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22BF4N
CAS Number:
Molecular Weight:
243.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

98.5-101.5% (T)
99%

form

powder

impurities

≤0.5% water

anion traces

bromide (Br-): ≤25 mg/kg
chloride (Cl-): ≤10 mg/kg
nitrate (NO3-): ≤25 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤25 mg/kg

SMILES string

F[B-](F)(F)F.CCCC[N+]1(C)CCCCC1

InChI

1S/C10H22N.BF4/c1-3-4-8-11(2)9-6-5-7-10-11;2-1(3,4)5/h3-10H2,1-2H3;/q+1;-1

InChI key

CGQPPCWBIGFWDL-UHFFFAOYSA-N

Application

1-Butyl-1-methylpiperidinium tetrafluoroborate is an ionic liquid, which can reduce the vulcanisation onset temperature during the vulcanisation of NBR (acrylonitrile-butadiene elastomer) composites.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yen-Hung Lin et al.
Science (New York, N.Y.), 369(6499), 96-102 (2020-07-08)
Longevity has been a long-standing concern for hybrid perovskite photovoltaics. We demonstrate high-resilience positive-intrinsic-negative perovskite solar cells by incorporating a piperidinium-based ionic compound into the formamidinium-cesium lead-trihalide perovskite absorber. With the bandgap tuned to be well suited for perovskite-on-silicon tandem
Effect of ionic liquids on the dispersion of zinc oxide and silica nanoparticles, vulcanisation behaviour and properties of NBR composites.
Maciejewska M and Zaborski M.
Express Polymer Letters, 8(12) (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service