Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

712957

Sigma-Aldrich

tert-Butyl (S)-2-pyrrolidone-5-carboxylate

≥98% (GC)

Synonym(s):

tert-Butyl (S)-5-oxo-2-pyrrolidinecarboxylate, tert-Butyl 5-oxo-L-prolinate, tert-Butyl L-pyroglutamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO3
CAS Number:
Molecular Weight:
185.22
Beilstein/REAXYS Number:
3590226
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥98% (GC)

form

powder

functional group

ester

SMILES string

CC(C)(C)OC(=O)[C@@H]1CCC(=O)N1

InChI

1S/C9H15NO3/c1-9(2,3)13-8(12)6-4-5-7(11)10-6/h6H,4-5H2,1-3H3,(H,10,11)/t6-/m0/s1

InChI key

QXGSPAGZWRTTOT-LURJTMIESA-N

Application

tert-Butyl (S)-2-pyrrolidone-5-carboxylate can be used:
  • As a starting material/synthon in the synthesis of angiotensin-converting enzyme inhibitors.[1][2]
  • In the synthesis of phenanthroindolizidine alkaloids named (+)-tylophorine and antofine.[3]
  • As a starting material in the synthesis of radioligands [18F]IUR-1602 and [18F]IUR-1601, applicable for imaging of P2X7R, a therapeutic target for neuroinflammation.[4][5]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and initial in vitro characterization of a new P2X7R radioligand [18F] IUR-1602
Gao M, et al.
Applied Radiation and Isotopes, 144(1), 10-18 (2019)
Indolizidine and quinolizidine alkaloids
Michael JP
Natural Product Reports, 24(1), 191-222 (2007)
Potent orally active inhibitors of angiotensin-converting enzyme (ACE)
Imaki K, et al.
Chemical & Pharmaceutical Bulletin, 29(8), 2210-2214 (1981)
Synthesis and preliminary biological evaluation of a novel P2X7R radioligand [18F] IUR-1601
Gao M, et al.
Bioorganic & medicinal chemistry letters, 28(9), 1603-1609 (2018)
Synthesis and pharmacology of the potent angiotensin-converting enzyme inhibitor N-[1 (S)-(ethoxycarbonyl)-3-phenylpropyl]-(S)-alanyl-(S)-pyroglutamic acid
Johnson AL, et al.
Journal of medicinal chemistry, 28(11), 1596-1602 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service