Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

712078

Sigma-Aldrich

(1S,2R)-(+)-2-Amino-1-cyclopentanecarboxylic acid hydrochloride hemihydrate

98.0-102.0% (calc. on dry substance, AT)

Synonym(s):

(+)-Cispentacin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2 · HCl · 0.5H2O
CAS Number:
Molecular Weight:
174.63
Beilstein/REAXYS Number:
627236
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

98.0-102.0% (calc. on dry substance, AT)

form

crystals

optical purity

enantiomeric excess: ≥98.0% (GC)

impurities

4.0-6.0% water

application(s)

peptide synthesis

SMILES string

O.Cl.Cl.N[C@@H]1CCC[C@@H]1C(O)=O.N[C@@H]2CCC[C@@H]2C(O)=O

InChI

1S/2C6H11NO2.2ClH.H2O/c2*7-5-3-1-2-4(5)6(8)9;;;/h2*4-5H,1-3,7H2,(H,8,9);2*1H;1H2/t2*4-,5+;;;/m00.../s1

InChI key

XAVWJWKIXZQILW-CNRKZEFXSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service