Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

711950

Sigma-Aldrich

(S)-3-(Boc-amino)-5-hexenoic acid

98%

Synonym(s):

Boc-(S)-3-amino-5-hexenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27
Beilstein/REAXYS Number:
9127734
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.5% (HPLC)
98%

form

lumps

optical activity

[α]/D +20±1°, c = 1 in ethanol

optical purity

enantiomeric excess: ≥99.0%

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CC=C)CC(O)=O

InChI

1S/C11H19NO4/c1-5-6-8(7-9(13)14)12-10(15)16-11(2,3)4/h5,8H,1,6-7H2,2-4H3,(H,12,15)(H,13,14)/t8-/m0/s1

InChI key

RFHPQLCVYMBPRF-QMMMGPOBSA-N

Application

(S)-3-(Boc-amino)-5-hexenoic acid can be used as a reactant for the preparation of lactam bridged peptide mimics by peptide coupling and olefin metathesis.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Short and efficient synthesis of homo-Freidinger lactams: an olefin metathesis approach towards conformationally restricted β-amino acid analogues
Hoffmann T and Gmeiner P
Synlett, 2002(06), 1014-1016 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service