Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

711705

Sigma-Aldrich

1-Propyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

≥98% (H-NMR)

Synonym(s):

PMIM BTA, PMIM TFSI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13F6N3O4S2
CAS Number:
Molecular Weight:
405.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98% (H-NMR)

form

liquid

impurities

≤0.5% water

SMILES string

CCCn1cc[n+](C)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C7H13N2.C2F6NO4S2/c1-3-4-9-6-5-8(2)7-9;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-7H,3-4H2,1-2H3;/q+1;-1

InChI key

CDWUIWLQQDTHRA-UHFFFAOYSA-N

General description

Sales restrictions may apply

Caution

not available in the US

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service