Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

711683

Sigma-Aldrich

1,4,7-Tri-Boc-1,4,7,10-tetraazacyclododecane

98%

Synonym(s):

N,N′,N′′-Tri-Boc-cyclen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H44N4O6
CAS Number:
Molecular Weight:
472.62
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

powder

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCNCCN(CCN(CC1)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C

InChI

1S/C23H44N4O6/c1-21(2,3)31-18(28)25-12-10-24-11-13-26(19(29)32-22(4,5)6)15-17-27(16-14-25)20(30)33-23(7,8)9/h24H,10-17H2,1-9H3

InChI key

ZXASHTYJQJMCQR-UHFFFAOYSA-N

Application

Building block used to attach the Cyclen azacrown to a molecular scaffold[1], building block for a trifold cyclen Zn complex as phosphate dianion receptor in aqueous solution[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimura, E., et al.
Journal of the American Chemical Society, 119, 3068-3068 (1997)
Chang Eun Yoo et al.
Journal of the American Chemical Society, 125(47), 14580-14589 (2003-11-20)
Mononuclear, dinuclear, and tetranuclear artificial metalloproteases were prepared by attaching respective catalytic modules containing the Cu(II) complex of cyclen (Cu(II)Cyc) to a derivative of cross-linked polystyrene. The polymeric artificial metalloproteases effectively cleaved peptide bonds of myoglobin (Mb) by hydrolysis. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service