Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

711519

Sigma-Aldrich

5-Trichloromethyl-4,5-dihydro-5-isoxazolol

≥97.0%

Synonym(s):

5-Hydroxy-5-trichloromethyl-4,5-dihydroisoxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4Cl3NO2
CAS Number:
Molecular Weight:
204.44
Beilstein/REAXYS Number:
2250086
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥97.0%
97.0-103.0% (T)

form

powder

mp

138-143 °C

SMILES string

OC1(CC=NO1)C(Cl)(Cl)Cl

InChI

1S/C4H4Cl3NO2/c5-4(6,7)3(9)1-2-8-10-3/h2,9H,1H2

InChI key

VHZMBXBPSPAPRH-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service