Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

711209

Sigma-Aldrich

Molybdenum(IV) oxide

97%

Synonym(s):

Molybdenum dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MoO2
CAS Number:
Molecular Weight:
127.94
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: molybdenum

density

6.47 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Mo]=O

InChI

1S/Mo.2O

InChI key

QXYJCZRRLLQGCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ding Zhou et al.
Nature communications, 5, 4716-4716 (2014-09-03)
Porous graphene, which features nano-scaled pores on the sheets, is mostly investigated by computational studies. The pores on the graphene sheets may contribute to the improved mass transfer and may show potential applications in many fields. To date, the preparation

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service