71106
Nω-(4-Methoxy-2,3,6-trimethylbenzenesulfonyl)-L-arginine-4-methoxybenzylamide hydrochloride
purum, ≥98.0% (HPLC)
Synonym(s):
N-(4-Methoxybenzyl)-Nω-(4-methoxy-2,3,6-trimethylbenzenesulfonyl)-L-argininamide hydrochloride
About This Item
grade
purum
assay
≥98.0% (HPLC)
form
crystals
reaction suitability
reaction type: solution phase peptide synthesis
application(s)
peptide synthesis
SMILES string
Cl.COc1ccc(CNC(=O)[C@@H](N)CCCNC(=N)NS(=O)(=O)c2c(C)cc(OC)c(C)c2C)cc1
InChI
1S/C24H35N5O5S.ClH/c1-15-13-21(34-5)16(2)17(3)22(15)35(31,32)29-24(26)27-12-6-7-20(25)23(30)28-14-18-8-10-19(33-4)11-9-18;/h8-11,13,20H,6-7,12,14,25H2,1-5H3,(H,28,30)(H3,26,27,29);1H/t20-;/m0./s1
InChI key
RXEUOACPUYTHGK-BDQAORGHSA-N
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service