Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

710350

Sigma-Aldrich

Sodium, 40 wt. % dispersion in oil

mixture of naphthenic oil and petroleum distillates

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na
Molecular Weight:
22.99
UNSPSC Code:
12141806

form

slurry

reaction suitability

core: sodium
reagent type: catalyst
reaction type: Reductions

reagent type: reductant

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Asp. Tox. 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - Water-react 1

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service