About This Item
assay
95%
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
109-113 °C
functional group
phosphine
SMILES string
COc1ccccc1P(Cl)c2ccccc2OC
InChI
1S/C14H14ClO2P/c1-16-11-7-3-5-9-13(11)18(15)14-10-6-4-8-12(14)17-2/h3-10H,1-2H3
InChI key
PEKLJSZEUFVLKO-UHFFFAOYSA-N
Application
- Cycloadditions reactions
- Asymmetric hydrogenation and allylic substitution
- Asymmetric hydrogenation of imines and olefins
Reactant for synthesis of:
- Ferrocene-based chiral diphosphine
- Catalysts for C-C cross coupling reactions
- Palladium complexes
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
supp_hazards
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service