Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

710148

Sigma-Aldrich

4-(Trifluoromethyl)phenylacetaldehyde dimethylacetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13F3O2
Molecular Weight:
234.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.448

density

1.158 g/mL at 25 °C

SMILES string

COC(Cc1ccc(cc1)C(F)(F)F)OC

InChI

1S/C11H13F3O2/c1-15-10(16-2)7-8-3-5-9(6-4-8)11(12,13)14/h3-6,10H,7H2,1-2H3

InChI key

LFPVFWUAVYTJNP-UHFFFAOYSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

194.9 °F

flash_point_c

90.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service