Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

709662

Sigma-Aldrich

α-(Boc-amino)phenethylamine

Synonym(s):

(2-Amino-1-phenyl-ethyl)carbamic acid tert-butyl ester, 1,1-Dimethylethyl (2-amino-1-phenylethyl)carbamate, 2-(Boc-amino)-2-phenylethanamine, tert-Butyl-(2-amino-1-phenylethyl)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$75.80

$75.80


Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
$75.80

About This Item

Empirical Formula (Hill Notation):
C13H20N2O2
CAS Number:
Molecular Weight:
236.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$75.80


Available to ship onApril 29, 2025Details


Request a Bulk Order

form

solid

Quality Level

mp

85-90 °C

functional group

amine
phenyl

SMILES string

CC(C)(C)OC(=O)NC(CN)c1ccccc1

InChI

1S/C13H20N2O2/c1-13(2,3)17-12(16)15-11(9-14)10-7-5-4-6-8-10/h4-8,11H,9,14H2,1-3H3,(H,15,16)

InChI key

IJALRZPKODHZOR-UHFFFAOYSA-N

Related Categories

Application

Reactant for:
  • Preparation of γ-lactams from carboxylic acids and amines via UDC (Ugi/De-BOC/Cyclize) strategy using isonitriles

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service