Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

709328

Sigma-Aldrich

3-[(Methylamino)methyl]pyridine

97%

Synonym(s):

N-Methyl-3-pyridinemethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.528

density

1.000 g/mL at 25 °C

functional group

amine

SMILES string

CNCc1cccnc1

InChI

1S/C7H10N2/c1-8-5-7-3-2-4-9-6-7/h2-4,6,8H,5H2,1H3

InChI key

MCSAQVGDZLPTBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

199.9 °F - closed cup

flash_point_c

93.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service