Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

709018

Sigma-Aldrich

Iron(III) arsenide

99.5% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeAs
CAS Number:
Molecular Weight:
130.77
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.5% trace metals basis

form

pieces

reaction suitability

reagent type: catalyst
core: arsenic

reagent type: catalyst
core: iron

mp

1020 °C

density

7.83 g/cm3 at 25 °C

SMILES string

[Fe]#[As]

InChI

1S/As.Fe

InChI key

VETKVGYBAMGARK-UHFFFAOYSA-N

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service