Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

708623

Sigma-Aldrich

(S)-Binam-P

95%

Synonym(s):

(S)-2,2′-Bis(diphenylphosphinoamino)-1,1′-binaphthyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H34N2P2
Molecular Weight:
652.70
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

optical activity

[α]22/D −31.0°, c = 0.5 in chloroform

mp

152-156 °C

SMILES string

N(P(c1ccccc1)c2ccccc2)c3ccc4ccccc4c3-c5c(NP(c6ccccc6)c7ccccc7)ccc8ccccc58

InChI

1S/C44H34N2P2/c1-5-19-35(20-6-1)47(36-21-7-2-8-22-36)45-41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46-48(37-23-9-3-10-24-37)38-25-11-4-12-26-38/h1-32,45-46H

InChI key

NRWZSRQDRSYPNW-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service