Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

708615

Sigma-Aldrich

(R)-Binam-P

Synonym(s):

(R)-2,2′-Bis(diphenylphosphinoamino)-1,1′-binaphthyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H34N2P2
CAS Number:
Molecular Weight:
652.70
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

solid

Quality Level

mp

151-156 °C

InChI

1S/C44H34N2P2/c1-5-19-35(20-6-1)47(36-21-7-2-8-22-36)45-41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46-48(37-23-9-3-10-24-37)38-25-11-4-12-26-38/h1-32,45-46H

InChI key

NRWZSRQDRSYPNW-UHFFFAOYSA-N

General description

(R)-Binam-P is an axially disymmetric bisphosphine ligand mainly used for rhodium-catalyzed asymmetric hydrogenation reactions.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(R)-2,2'-Bis(diphenylphosphinoamino)-1,1'-binaphthyl.
Schneider JF.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service