Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

708585

Sigma-Aldrich

3-Mesityl-1-methyl-1H-benzimidazolium iodide

95%

Synonym(s):

1-Methyl-3-(2,4,6-trimethylphenyl)-1H-benzimidazolium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19IN2
CAS Number:
Molecular Weight:
378.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

95%

form

solid

reaction suitability

reagent type: catalyst

mp

267-273 °C

SMILES string

[I-].Cc1cc(C)c(c(C)c1)-n2c[n+](C)c3ccccc23

InChI

1S/C17H19N2.HI/c1-12-9-13(2)17(14(3)10-12)19-11-18(4)15-7-5-6-8-16(15)19;/h5-11H,1-4H3;1H/q+1;/p-1

InChI key

GZUMUMKCXFZBLQ-UHFFFAOYSA-M

Application

Reagent for β-protonations of homoenolate equivalents

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service