Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

708151

Sigma-Aldrich

5-(Trifluoromethyl)pyridine-3-carbonitrile

95%

Synonym(s):

5-(Trifluoromethyl)nicotinonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$19.18

$19.18

List Price$27.40Save 30%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
$19.18

About This Item

Empirical Formula (Hill Notation):
C7H3F3N2
CAS Number:
Molecular Weight:
172.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$19.18

List Price$27.40Save 30%
Web-Only Promotion

Available to ship onMay 01, 2025Details


Request a Bulk Order

Quality Level

assay

95%

form

solid

mp

42-46 °C

functional group

fluoro
nitrile

SMILES string

FC(F)(F)c1cncc(c1)C#N

InChI

1S/C7H3F3N2/c8-7(9,10)6-1-5(2-11)3-12-4-6/h1,3-4H

Inchi Key

HAWFNLPVIMFXGB-UHFFFAOYSA-N

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

180.0 °F

flash_point_c

82.2 °C

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service