Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

707988

Sigma-Aldrich

Grubbs Catalyst® C859

95%

Synonym(s):

Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene][(tricyclohexylphosphoranyl)methylidene]ruthenium(II) tetrafluoroborate, Piers second generation metathesis catalyst, [SIMes]dichloro[(tricyclohexylphosphoranyl)methylidene]Ru tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H60BCl2F4N2PRu
CAS Number:
Molecular Weight:
858.67
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Ring-Opening Polymerization

mp

176 °C (dec.)

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.Cc1cc(C)c(N2CCN(c3c(C)cc(C)cc3C)C2=[Ru](Cl)(Cl)=C[P+](C4CCCCC4)(C5CCCCC5)C6CCCCC6)c(C)c1

InChI

1S/C21H26N2.C19H34P.BF4.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;2-1(3,4)5;;;/h9-12H,7-8H2,1-6H3;1,17-19H,2-16H2;;2*1H;/q;+1;-1;;;+2/p-2

InChI key

KMTPXIJXNIAPCS-UHFFFAOYSA-L

Application

Active as low as -50 °C, which enables direct observation of reaction intermediates.

Legal Information

Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapidly initiating ruthenium olefin-metathesis catalysts.
Romero PE, et al.
Angewandte Chemie (International Edition in English), 43(45), 6161-6165 (2004)
Kinetic and thermodynamic analysis of processes relevant to initiation of olefin metathesis by ruthenium phosphonium alkylidene catalysts.
Leitao EM, et al.
Journal of the American Chemical Society, 132(8), 2784-2794 (2010)

Articles

Alkyne metathesis has been a useful tool for C–C bond formation since the discovery of structurally well-defined metal alkylidynes by Schrock and coworkers.

Related Content

Research focuses on ruthenium alkylidene complexes for versatile olefin metatheses catalysis in the Grubbs group.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service