Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

707872

Sigma-Aldrich

2,6-Dimethylphenylboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16BNO4
Molecular Weight:
261.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

217-222 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2c(C)cccc2C

InChI

1S/C13H16BNO4/c1-9-5-4-6-10(2)13(9)14-18-11(16)7-15(3)8-12(17)19-14/h4-6H,7-8H2,1-3H3

InChI key

DFYUBULBBZGRLF-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service