707287
Allyl-13C3-amine-15N hydrochloride
99 atom % 13C, 98 atom % 15N, 98% (CP)
Synonym(s):
3-Aminopropene-13C3,15N hydrochloride
About This Item
Recommended Products
isotopic purity
99 atom % 13C
98 atom % 15N
assay
98% (CP)
form
powder
mp
106 °C
mass shift
M+4
SMILES string
Cl.[15NH2][13CH2][13CH]=[13CH2]
InChI
1S/C3H7N.ClH/c1-2-3-4;/h2H,1,3-4H2;1H/i1+1,2+1,3+1,4+1;
Inchi Key
MLGWTHRHHANFCC-UJNKEPEOSA-N
Related Categories
Packaging
signalword
Danger
hcodes
pcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service