Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

706671

Sigma-Aldrich

(4S,5S)-2-Allyl-2-chloro-3,4-dimethyl-5-phenyl-1-oxa-3-aza-2-silacyclopentane

Synonym(s):

(1S,2S)-pseudoephedrine allylchlorosilane, (4S,5S)-2-Allyl-2-chloro-3,4-dimethyl-5-phenyl-1,3,2-oxazasilolidine, (4S,5S)-2-Chloro-3,4-dimethyl-5-phenyl-2-(2-propenyl)-1-oxa-3-aza-2-silacyclopentane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18ClNOSi
CAS Number:
Molecular Weight:
267.83
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

liquid

optical activity

[α]20/D 72.5°, c = 0.6 in ethanol (200 proof)

refractive index

n20/D 1.517

bp

120 °C/5 mmHg

density

1.095 g/mL at 25 °C

SMILES string

C[C@H]1[C@@H](O[Si](Cl)(CC=C)N1C)c2ccccc2

InChI

1S/C13H18ClNOSi/c1-4-10-17(14)15(3)11(2)13(16-17)12-8-6-5-7-9-12/h4-9,11,13H,1,10H2,2-3H3/t11-,13+,17?/m0/s1

InChI key

XHCRAANIMMYPDV-APBZJUGRSA-N

General description

This product is a mixture of diastereomers.

Application

Leighton′s Strained Silacycles: Powerful Enantioselective Allylation Reagents

  • Practical, enantioselective allylation of acylhydrazones
  • Enantioselective allylation of ketimines: synthesis of tertiary carbinamines
Reagent used in the enantioselective allylation of aldehydes

Legal Information

U.S. Patent No. 7,534,905

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

target_organs

Respiratory system

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

100.4 °F

flash_point_c

38 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James W A Kinnaird et al.
Journal of the American Chemical Society, 124(27), 7920-7921 (2002-07-04)
A new reagent for the enantioselective allylation of aliphatic aldehydes has been developed. The reagent is easily prepared in a single step from commercially available materials and may be stored without significant decomposition. The reactivity of the reagent is attributed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service