Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

706450

Sigma-Aldrich

(R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether solution

0.05 M in toluene

Synonym(s):

(R)-α,α-Diphenylprolinol trimethylsilyl ether solution, (R)-2-[Diphenyl(trimethylsilanyloxy)methyl]pyrrolidine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27NOSi
Molecular Weight:
325.52
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

0.05 M in toluene

refractive index

n20/D 1.498

density

0.865 g/mL at 25 °C

functional group

phenyl

SMILES string

C[Si](C)(C)OC([C@H]1CCCN1)(c2ccccc2)c3ccccc3

InChI

1S/C20H27NOSi/c1-23(2,3)22-20(19-15-10-16-21-19,17-11-6-4-7-12-17)18-13-8-5-9-14-18/h4-9,11-14,19,21H,10,15-16H2,1-3H3/t19-/m1/s1

InChI key

RSUHWMSTWSSNOW-LJQANCHMSA-N

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F

flash_point_c

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service