Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

706051

Sigma-Aldrich

1-Propyl-1H-pyrazole-4-boronic acid pinacol ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21BN2O2
CAS Number:
Molecular Weight:
236.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.476

density

0.996 g/mL at 25 °C

SMILES string

CCCn1cc(cn1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C12H21BN2O2/c1-6-7-15-9-10(8-14-15)13-16-11(2,3)12(4,5)17-13/h8-9H,6-7H2,1-5H3

Inchi Key

BKLGYJWLZWMIDO-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

109.9 °F

flash_point_c

43.3 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service