Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

706000

Sigma-Aldrich

Fluorotris(triphenylphosphine)copper(I)

95% (by HMR)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H45CuFP3
CAS Number:
Molecular Weight:
869.40
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

95% (by HMR)

form

solid

reaction suitability

reagent type: catalyst
core: copper

mp

144-157 °C

SMILES string

F[Cu].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9

InChI

1S/3C18H15P.Cu.FH/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h3*1-15H;;1H/q;;;+1;/p-1

Inchi Key

OKJARODAQSGFQE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Product may contain up to 9 wt. % ethanol as a complex.

Application

Complex used in asymmetric aldol reaction[1]

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kounosuke Oisaki et al.
Journal of the American Chemical Society, 128(22), 7164-7165 (2006-06-01)
An enantioselective aldol reaction between ketones and ketene silyl acetals is described using CuF-chiral phosphine as a catalyst. The key for high enantioselectivity was the development of a novel ligand derived from Taniaphos combined with the unique accelerative effect of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service