705284
1,1′-Carbonyldiimidazole solution
0.4 M in methylene chloride
About This Item
Recommended Products
form
liquid
reaction suitability
reaction type: Carbonylations
concentration
0.4 M in methylene chloride
refractive index
n20/D 1.434
density
1.303 g/mL at 25 °C
SMILES string
O=C(n1ccnc1)n2ccnc2
InChI
1S/C7H6N4O/c12-7(10-3-1-8-5-10)11-4-2-9-6-11/h1-6H
InChI key
PFKFTWBEEFSNDU-UHFFFAOYSA-N
General description
Application
- Synthesis of peptides in aqueous solution.[2]
- Synthesis of isoleucine-5 angiotensin II amide-1.[1]
- For the activation of the hydroxyl termini of tween toward nucleophilic addition.[3]
- As coupling reagent for the coupling of prostaglandins and other carboxylic acids to the amino groups of bovine serum albumin.[4]
- Synthesis of peptide thioesters.[5]
signalword
Danger
hcodes
Hazard Classifications
Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
target_organs
Central nervous system
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service