Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

704644

Sigma-Aldrich

N-(Trimethylsilyl)-4-chlorobenzaldimine

95%

Synonym(s):

N-(4-Chlorobenzylidene)trimethylsilylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14ClNSi
CAS Number:
Molecular Weight:
211.76
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

form

solid

mp

27-36 °C

SMILES string

C[Si](C)(C)\N=C\c1ccc(Cl)cc1

InChI

1S/C10H14ClNSi/c1-13(2,3)12-8-9-4-6-10(11)7-5-9/h4-8H,1-3H3/b12-8+

InChI key

XLCRIROPKDZGLD-XYOKQWHBSA-N

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 2

flash_point_f

129.9 °F

flash_point_c

54.4 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service