Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

704636

Sigma-Aldrich

Methyl indole-1-carboxylate

97%

Synonym(s):

N-(Methoxycarbonyl)indole, Indole-1-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

liquid

refractive index

n20/D 1.583

density

1.191 g/mL at 25 °C

SMILES string

COC(=O)n1ccc2ccccc12

InChI

1S/C10H9NO2/c1-13-10(12)11-7-6-8-4-2-3-5-9(8)11/h2-7H,1H3

InChI key

FHOPBCYBMWVMGJ-UHFFFAOYSA-N

Application

  • Reactant for preparation of Bcl Inhibitor Obatoclax
  • Reactant for preparation of tetracyclic indolines
  • Reactant for Friedel-Crafts alkylation reaction of indoles
  • Reactant for oxidative free radical reactions

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

>212.0 °F

flash_point_c

> 100 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service