Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

704512

Sigma-Aldrich

Strontium acetylacetonate hydrate

Synonym(s):

Sr(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3COCH=C(O-)CH3]2Sr · xH2O
CAS Number:
Molecular Weight:
285.84
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Sr, 29.6-31.8% EDTA titration (dry basis)

reaction suitability

core: strontium
reagent type: catalyst

mp

>300

SMILES string

CC(=O)\C=C(\C)O[Sr]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Sr/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

UMBFGWVRZIHXCK-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service