Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

704253

Sigma-Aldrich

Lanthanum strontium cobalt ferrite

greener alternative

composite cathode powder, LSCF/GDC

Synonym(s):

LCSF, Lanthanum strontium cobalt iron oxide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

form

powder

composition

Ce0.9Gd0.102, 50 wt. %
La0.6Sr0.4Co0.2Fe0.8O3 , 50 wt. %

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

0.4-0.9 μm (Horiba-centrifugal)

application(s)

battery manufacturing

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Lanthanum strontium cobalt ferrite (LSCF) is a mixed ionic and electron conducting pervoskite. The ionic conductivity is lower than YSZ although maintaining high conductivity.[1] X-ray diffraction and electrochemical measurements of an operational LSCF cathode was conducted to study its lattice expansion during use.[2] LSCF can be prepared by solid state reaction of individual cation oxides.[3]
Lanthanum strontium cobalt ferrite (LSCF/GDC) belongs to the class of cathode materials that operate at low temperatures. It has mixed ionic and electronic conductivities with high electrocatalytic activity. It forms thin films with higher performance than the thick dense film.[4]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

LSCF may be used as cathode in solid oxide fuel cells (SOFCs). [5]
LSCF oxygen electrodes can be used in the fabrication of high-performance solid oxide fuel cells (SOFCs).[4][6][7][8]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low-temperature electrochemical characterization of dense ultra-thin lanthanum strontium cobalt ferrite (La0. 6Sr0. 4Co0. 8Fe0. 2O3) cathodes synthesized by RF-sputtering on nanoporous alumina-supported Y-doped zirconia membranes
Xiong H, et al.
Journal of Power Sources, 193(2), 589-592 (2009)
Electrochemical performance and stability of lanthanum strontium cobalt ferrite oxygen electrode with gadolinia doped ceria barrier layer for reversible solid oxide fuel cell
Fan H, et al.
Journal of Power Sources, 268, 634-639 (2014)
Preparation of an asymmetric perovskite-type membrane and its oxygen permeability
Jin W, et al.
Journal of Membrane Science, 185(2), 237-243 (2001)
Advances in Materials for Solid Oxide Fuel Cells
Gorte RJ, et al.
Material Matters, 5(4) null
Lattice expansion of LSCF-6428 cathodes measured by in situ XRD during SOFC operation.
Hardy JS, et al.
Journal of Power Sources, 198, 76-82 (2012)

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Questions

1–5 of 5 Questions  
  1. What are the benefits of Product 704253, Lanthanum strontium cobalt - composite cathode powder?

    1 answer
    1. Composite cathodes are used for their enhanced catalytic activity.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. How do I apply Product 704253, Lanthanum strontium cobalt - composite cathode powder, onto electrolyte substrates?

    1 answer
    1. This material is readily mixed into an ink for coating onto electrolyte substrates, or tape casting is possible as well.

      Helpful?

  4. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  5. What electrolytes can Product 704253, Lanthanum strontium cobalt - composite cathode powder, be used with?

    1 answer
    1. This cathode material can be used with either ceria- or zirconia-based electrolytes.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service