Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

704210

Sigma-Aldrich

Nickel oxide - Cerium samarium oxide

greener alternative

for coatings, NiO/SDC, ≥99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

assay

≥99% trace metals basis

form

solid

composition

(CeO2-Sm2O3), 40 wt. %
NiO, 60 wt. %

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

89-90 °C

application(s)

battery manufacturing

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

NiO-SDC composite shows a high catalytic activity as an anode material, which makes it a potential candidate for the fabrication of solid oxide fuel cells (SOFCs). It shows a maximum power density of ~490 mW cm-2 at 600°C and a polarization resistance of 0.1 Ωcm2.[1][2][3][4]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 4 - Carc. 1A Inhalation - Skin Sens. 1 - STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lanthanum promoted NiO-SDC anode for low temperature solid oxide fuel cells fueled with methane
Yan A, et al.
Journal of Power Sources, 210(2), 374-380 (2012)
Electrochemical performance of gel-cast NiO-SDC composite anodes in low-temperature SOFCs
Yin Y, et al.
Electrochimica Acta, 51(13), 2594-2598 (2006)
Ni-SDC cermet anode fabricated from NiO-SDC composite powder for intermediate temperature SOFC
Misono T, et al.
Journal of Power Sources, 157(2), 754-757 (2006)
Gel-cast NiO-SDC composites as anodes for solid oxide fuel cells
Yin Y, et al.
Journal of Power Sources, 132(1-2), 36-41 (2004)

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Questions

1–5 of 5 Questions  
  1. What is the surface area of Product 704210, Nickel oxide - Cerium samarium oxide?

    1 answer
    1. According to our supplier, the surface area is 4-8 m2/g.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. What is the particle size of Product 704210, Nickel oxide - Cerium samarium oxide?

    1 answer
    1. This material is a multi-component mixture, so there are many different particle size ranges in the material.

      Helpful?

  4. What firing temperatures are needed for Product 704210, Nickel oxide - Cerium samarium oxide, when making an electrode?

    1 answer
    1. According to our supplier, firing temperatures are on the order of 1200-1350°C.

      Helpful?

  5. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service