Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

701904

Sigma-Aldrich

Methyl 2-fluoropyridine-3-carboxylate

97%

Synonym(s):

2-Fluoronicotinic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6FNO2
CAS Number:
Molecular Weight:
155.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

liquid

bp

75 °C/5 mmHg

mp

22-26 °C

functional group

ester
fluoro

SMILES string

COC(=O)c1cccnc1F

InChI

1S/C7H6FNO2/c1-11-7(10)5-3-2-4-9-6(5)8/h2-4H,1H3

InChI key

PJNHJXKXBSOLBH-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F

flash_point_c

110 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service